Merck
  • Beta-adrenergic receptors are differentially expressed in distinct interneuron subtypes in the rat hippocampus.
Beta-adrenergic receptors are differentially expressed in distinct interneuron subtypes in the rat hippocampus.
The Journal of comparative neurology (2008-06-12)
David J Cox, Claudia Racca, Fiona E N LeBeau
ABSTRACT
Noradrenaline (NA) acting via beta-adrenergic receptors (betaARs) plays an important role in the modulation of memory in the hippocampus. betaARs have been shown to be expressed in principal cells, but their distribution across different interneuron classes is unknown. We have used specific interneuron markers including calcium binding proteins (parvalbumin, calbindin, and calretinin) and neuropeptides (somatostatin, neuropeptide Y, and cholecystokinin) together with either beta1AR or beta2AR to determine the distribution of these receptors in all major subfields of the hippocampus. We found that beta1AR-expressing interneurons were more prevalent in the CA3 and CA1 regions of the hippocampus than in the dentate gyrus, where they were relatively sparse. beta2AR-expressing interneurons were more uniformly distributed between all three regions of the hippocampus. A high proportion of neuropeptide Y-containing interneurons in the dentate gyrus co-expressed beta2AR. beta1AR labeling was common in interneurons expressing somatostatin and parvalbumin in the CA3 and CA1 regions, particularly in the stratum oriens of these regions. beta2AR labeling was more likely to be found than beta1AR labeling in cholecystokinin-expressing interneurons. In contrast, calretinin-containing interneurons were virtually devoid of beta1AR or beta2AR labeling. These regional and interneuron type-specific differences suggest functionally distinct roles for NA in modulating hippocampal activity via activation of betaARs.
Sigma-Aldrich
Anti-Neuropeptide Y Antibody, serum, Chemicon®