MilliporeSigma
  • In vivo and in vitro action of new antibiotics interfering with the utilization of N-acetyl-glucosamine-N-acetyl-muramyl-pentapeptide.
In vivo and in vitro action of new antibiotics interfering with the utilization of N-acetyl-glucosamine-N-acetyl-muramyl-pentapeptide.
Journal of bacteriology (1971-10-01)
E J Lugtenberg, A v Schijndel-van Dam, T H van Bellegem
ABSTRACT
Recent literature on the antibiotics enduracidin, moenomycin, prasinomycin, and 11.837 RP suggested an interaction with murein synthesis. Incubation of sensitive strains from Bacillus cereus and Staphylococcus aureus in a "wall medium" containing labeled l-alanine showed that all four antibiotics inhibited the incorporation of alanine into murein and gave rise to accumulation of radioactive uridine diphosphate-N-acetyl-muramyl (UDP-MurNAc)-pentapeptide. Peptidoglycan was synthesized when the particulate enzyme of B. stearothermophilus was incubated with the murein precursors UDP-N-acetyl-glucosamine (UDP-GlcNAc) and UDP-MurNAc-pentapeptide. The newly formed polymer was less accessible for lysozyme and more strongly bound to the acceptor than the same product from the Escherichia coli particulate enzyme. After incubation in the presence of penicillin, a greater part of the peptidoglycan was lysozyme sensitive and more loosely bound to the acceptor. The antibiotics enduracidin, moenomycin, prasinomycin, and 11.837 RP inhibited peptidoglycan synthesis by the B. stearothermophilus particulate enzyme. The rate of synthesis of GlcNAc-MurNAc(-pentapeptide)-P-P-phospholipid was independent from the addition of these antibiotics, but its utilization was strongly inhibited. With the present results, it is not possible to distinguish the mechanisms of action of enduracidin, moenomycin, prasinomycin, and 11.837 RP from the mechanisms of action of vancomycin and ristocetin.