Merck
  • Establishment and characterization of new human embryonic stem cell lines.
Establishment and characterization of new human embryonic stem cell lines.
Reproductive biomedicine online (2005-06-14)
Necati Findikli, Semra Kahraman, Oya Akcin, Semra Sertyel, Zafer Candan
ABSTRACT
Human embryonic stem cells (hESC), with their ability to differentiate into all cell types in the human body, are likely to play a very important therapeutic role in a variety of neurodegenerative and life-threatening disorders in the near future. Although more than 120 different human embryonic stem cell lines have been reported worldwide, only a handful are currently available for researchers, which limits the number of studies that can be performed. This study reports the isolation, establishment and characterization of new human embryonic stem cell lines, as well as their differentiation potential into variety of somatic cell types. Blastocyst-stage embryos donated for research after assisted reproductive techniques were used for embryonic stem cell isolation. A total of 31 blastocysts were processed either for immunosurgery or direct culture methods for inner cell mass isolation. A total of nine primary stem cell colonies were isolated and of these, seven cell lines were further expanded and passaged. Established lines were characterized by their cellular and colony morphology, karyotypes and immunocytochemical properties. They were also successfully cryopreserved/thawed and showed similar growth and cellular properties upon thawing. When induced to differentiate in vitro, these cells formed a variety of somatic cell lineages including cells of endoderm, ectoderm and mesoderm origin. There is now an exponentially growing interest in stem cell biology as well as its therapeutic applications for life-threatening human diseases. However, limited availability of stem cell lines as well as financial or ethical limitations restrict the number of research projects. The establishment of new hESC lines may create additional potential sources for further worldwide and nationwide research on stem cells.
Sigma-Aldrich
ES Cell Characterization Kit, The Embryonic Stem Cell Characterization Kit phenotypically assesses the differentiation status of ES cells by measuring their AP activity, cell-surface stage-specific antigens (SSEA-1, SSEA-4) as well as expression of TRA-1-60, TRA-1-81 antigens.
Sigma-Aldrich
Leukemia Inhibitory Factor human, 5 µg, human recombinant LIF protein, expressed in E. coli, suitable for stem cell culture
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse