Merck
Mechanisms of IL-10 production in human microglia-T cell interaction.
Journal of immunology (Baltimore, Md. : 1950) (1999-06-03)
S Chabot, G Williams, M Hamilton, G Sutherland, V W Yong
ABSTRACT
IL-10, a cytokine with important anti-inflammatory properties, is generated within the CNS during neuroinflammation. The mechanism for its production is poorly understood. Since infiltrating lymphocytes come into close proximity with the macrophage-like cells of the CNS, the microglia, we have used an in vitro human microglia-T cell coculture system to address the mechanisms of IL-10 production. We demonstrate that microglia or activated T cells alone secrete negligible amounts of IL-10, but that their coculture results in significant IL-10 production, which was effected by both cell types. IL-10 generation was cell contact dependent, and treatment with anti-CD40, CTLA-4-Fc, or anti-CD23 decreased the IL-10 content in microglia-T cell cocultures. The combination of anti-CD40 and CTLA-4-Fc reduced IL-10 levels to the negligible amounts seen with T cells or microglia in isolation. By also measuring TNF-alpha levels, specificity of cytokine regulation was observed; while anti-CD40 and CTLA-4-Fc reduced IL-10 and TNF-alpha levels, anti-CD23 did not affect TNF-alpha while attenuating IL-10 generation. Anti-very late Ag-4, which decreased TNF-alpha levels, did not affect IL-10. These results implicate the CD40, B7, and CD23 pathways in IL-10 production following microglia-T cell encounter and have relevance to the regulation of an anti-inflammatory response within the CNS.
Sigma-Aldrich
IgG1 Antibody, Mouse, 1.95 mg/mL, Chemicon®