Merck
  • Effect of Roasting on Oligosaccharide Abundance in Arabica Coffee Beans.
Effect of Roasting on Oligosaccharide Abundance in Arabica Coffee Beans.
Journal of agricultural and food chemistry (2018-09-04)
Tian Tian, Samara Freeman, Mark Corey, J Bruce German, Daniela Barile
ABSTRACT
Emerging research into the bioactivities of indigestible carbohydrates is illuminating the potential of various foods and food streams to serve as novel sources of health-promoting compounds. Oligosaccharides (OS) are widely present in milks and some plants. Our previous research demonstrated the presence of OS in brewed coffee and spent coffee grounds. Armed with this new knowledge, the next step toward improving the utilization of these valuable components involved investigating the effect of roasting on the formation and abundance of coffee OS. In the present study, we used advanced mass spectrometry to analyze a variety of coffee samples and demonstrated that a great structural diversity and increased abundance of OS is associated with higher roasting intensity. The present investigation also evaluated methods for OS extraction and fractionation. A preparative-scale chromatographic method, based on activated carbon, was developed to isolate enough amounts of OS from coffee to enable future confirmation of prebiotic and other in vitro activities.
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG