Merck
  • Chromatic summation and receptive field properties of blue-on and blue-off cells in marmoset lateral geniculate nucleus.
Chromatic summation and receptive field properties of blue-on and blue-off cells in marmoset lateral geniculate nucleus.
Vision research (2017-11-14)
C D Eiber, A N J Pietersen, N Zeater, S G Solomon, P R Martin
ABSTRACT
The "blue-on" and "blue-off" receptive fields in retina and dorsal lateral geniculate nucleus (LGN) of diurnal primates combine signals from short-wavelength sensitive (S) cone photoreceptors with signals from medium/long wavelength sensitive (ML) photoreceptors. Three questions about this combination remain unresolved. Firstly, is the combination of S and ML signals in these cells linear or non-linear? Secondly, how does the timing of S and ML inputs to these cells influence their responses? Thirdly, is there spatial antagonism within S and ML subunits of the receptive field of these cells? We measured contrast sensitivity and spatial frequency tuning for four types of drifting sine gratings: S cone isolating, ML cone isolating, achromatic (S + ML), and counterphase chromatic (S - ML), in extracellular recordings from LGN of marmoset monkeys. We found that responses to stimuli which modulate both S and ML cones are well predicted by a linear sum of S and ML signals, followed by a saturating contrast-response relation. Differences in sensitivity and timing (i.e. vector combination) between S and ML inputs are needed to explain the amplitude and phase of responses to achromatic (S + ML) and counterphase chromatic (S - ML) stimuli. Best-fit spatial receptive fields for S and/or ML subunits in most cells (>80%) required antagonistic surrounds, usually in the S subunit. The surrounds were however generally weak and had little influence on spatial tuning. The sensitivity and size of S and ML subunits were correlated on a cell-by-cell basis, adding to evidence that blue-on and blue-off receptive fields are specialised to signal chromatic but not spatial contrast.
Sigma-Aldrich
Bromoenol lactone, ≥98% (TLC)