Merck
RPGRIP1L helps to establish the ciliary gate for entry of proteins.
Journal of cell science (2018-09-22)
Huawen Lin, Suyang Guo, Susan K Dutcher
ABSTRACT
Mutations in transition zone genes change the composition of the ciliary proteome. We isolated new mutations in RPGRIP1L (denotated as RPG1 in algae) that affect the localization of the transition zone protein NPHP4 in the model organism Chlamydomonas reinhardtii NPHP4 localization is not affected in multiple new intraflagellar transport (IFT) mutants. We compared the proteome of cilia from wild-type and mutants that affect the transition zone (RPGRIP1L) or IFT (IFT172 and DHC1b) by mass spectrometry. The rpg1-1 mutant cilia show the most dramatic increase in cytoplasmic proteins. These nonciliary proteins function in translation, membrane remodeling, ATP production and as chaperonins. These proteins are excluded in isolated cilia from fla11-1 (IFT172) and fla24-1 (DHC1b). Our data support the idea that RPGRIP1L, but not IFT proteins, acts as part of the gate for cytoplasmic proteins. The rpg1-1 cilia lack only a few proteins, which suggests that RPGRIP1L only has a minor role of in the retention of ciliary proteins. The fla11-1 mutant shows the greatest loss/reduction of proteins, and one-third of these proteins have a transmembrane domain. Hence, IFT172 may play a role in the retention of proteins.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® antibody produced in rabbit, clone SIG1-25, ascites fluid
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-TCP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture