Merck
  • microRNA-200b modulates microglia-mediated neuroinflammation via the cJun/MAPK pathway.
microRNA-200b modulates microglia-mediated neuroinflammation via the cJun/MAPK pathway.
Journal of neurochemistry (2014-04-23)
Shweta P Jadhav, Sandhya P Kamath, Mahesh Choolani, Jia Lu, S Thameem Dheen
ABSTRACT
Chronic activation of microglia, the macrophages of the CNS, has been shown to enhance neuronal damage because of excessive release of proinflammatory cytokines and neurotoxic molecules in a number of neurodegenerative diseases. Recent reports showed altered microRNA (miRNA) expression in immune-mediated pathologies, thus suggesting that miRNAs modulate expression of genes involving immune responses. This study demonstrates that miRNA-200b is expressed in microglia and modulates inflammatory response of microglia by regulating mitogen-activated protein kinase pathway. miRNA-200b expression was found to be down-regulated in activated microglia in vivo (traumatic brain injury rat model) and in vitro. A luciferase assay and loss- and gain-of-function studies revealed c-Jun, the transcription factor of cJun-N terminal kinase (JNK) mitogen-activated protein kinase pathway to be the target of miR-200b. Knockdown of miR-200b in microglia increased JNK activity along with an increase in pro-inflammatory cytokines, inducible nitric oxide synthase expression and nitric oxide (NO) production. Conversely, over-expression of miRNA-200b in microglia resulted in a decrease in JNK activity, inducible nitric oxide synthase expression, NO production and migratory potential of activated microglia. Furthermore, miR-200b inhibition resulted in increased neuronal apoptosis after treatment of neuronal cells with conditioned medium obtained from microglial culture. Taken together, these results indicate that miRNA-200b modulates microglial inflammatory process including cytokine secretion, NO production, migration and neuronal survival.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Chemicon®, from goat
Millipore
Nuclear Extraction Kit
Sigma-Aldrich
Anti-Tumor Necrosis Factor-α Antibody, Chemicon®, from rabbit