Merck
  • Prostanoid EP₂ Receptors Are Up-Regulated in Human Pulmonary Arterial Hypertension: A Key Anti-Proliferative Target for Treprostinil in Smooth Muscle Cells.
Prostanoid EP₂ Receptors Are Up-Regulated in Human Pulmonary Arterial Hypertension: A Key Anti-Proliferative Target for Treprostinil in Smooth Muscle Cells.
International journal of molecular sciences (2018-08-15)
Jigisha A Patel, Lei Shen, Susan M Hall, Chabha Benyahia, Xavier Norel, Robin J McAnulty, Shahin Moledina, Adam M Silverstein, Brendan J Whittle, Lucie H Clapp
ABSTRACT
Prostacyclins are extensively used to treat pulmonary arterial hypertension (PAH), a life-threatening disease involving the progressive thickening of small pulmonary arteries. Although these agents are considered to act therapeutically via the prostanoid IP receptor, treprostinil is the only prostacyclin mimetic that potently binds to the prostanoid EP₂ receptor, the role of which is unknown in PAH. We hypothesised that EP₂ receptors contribute to the anti-proliferative effects of treprostinil in human pulmonary arterial smooth muscle cells (PASMCs), contrasting with selexipag, a non-prostanoid selective IP agonist. Human PASMCs from PAH patients were used to assess prostanoid receptor expression, cell proliferation, and cyclic adenosine monophosphate (cAMP) levels following the addition of agonists, antagonists or EP₂ receptor small interfering RNAs (siRNAs). Immunohistochemical staining was performed in lung sections from control and PAH patients. We demonstrate using selective IP (RO1138452) and EP₂ (PF-04418948) antagonists that the anti-proliferative actions of treprostinil depend largely on EP₂ receptors rather than IP receptors, unlike MRE-269 (selexipag-active metabolite). Likewise, EP₂ receptor knockdown selectively reduced the functional responses to treprostinil but not MRE-269. Furthermore, EP₂ receptor levels were enhanced in human PASMCs and in lung sections from PAH patients compared to controls. Thus, EP₂ receptors represent a novel therapeutic target for treprostinil, highlighting key pharmacological differences between prostacyclin mimetics used in PAH.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human PTGER2
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
N-Octanoyl-N-methylglucamine, ≥97% (GC)