Merck
  • Notch-inducing hydrogels reveal a perivascular switch of mesenchymal stem cell fate.
Notch-inducing hydrogels reveal a perivascular switch of mesenchymal stem cell fate.
EMBO reports (2018-07-04)
Ulrich Blache, Queralt Vallmajo-Martin, Edward R Horton, Julien Guerrero, Valentin Djonov, Arnaud Scherberich, Janine T Erler, Ivan Martin, Jess G Snedeker, Vincent Milleret, Martin Ehrbar
ABSTRACT
The fate of mesenchymal stem cells (MSCs) in the perivascular niche, as well as factors controlling their fate, is poorly understood. Here, we study MSCs in the perivascular microenvironment of endothelial capillaries by modifying a synthetic 3D biomimetic poly(ethylene glycol) (PEG)-hydrogel system in vitro We show that MSCs together with endothelial cells form micro-capillary networks specifically in soft PEG hydrogels. Transcriptome analysis of human MSCs isolated from engineered capillaries shows a prominent switch in extracellular matrix (ECM) production. We demonstrate that the ECM phenotypic switch of MSCs can be recapitulated in the absence of endothelial cells by functionalizing PEG hydrogels with the Notch-activator Jagged1. Moreover, transient culture of MSCs in Notch-inducing microenvironments reveals the reversibility of this ECM switch. These findings provide insight into the perivascular commitment of MSCs by use of engineered niche-mimicking synthetic hydrogels.
Sigma-Aldrich
MISSION® esiRNA, targeting human FN1
Sigma-Aldrich
Decamethylcyclopentasiloxane, 97%
Sigma-Aldrich
GM 6001, InSolution, ≥95%, broad-spectrum inhibitor of MMPs
Sigma-Aldrich
MMP-2 Inhibitor III, The MMP-2 Inhibitor III, also referenced under CAS 704888-90-4, controls the biological activity of MMP-2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.