Merck
  • Genetic variation in IGF1, IGFBP3, IRS1, IRS2 and risk of breast cancer in women living in Southwestern United States.
Genetic variation in IGF1, IGFBP3, IRS1, IRS2 and risk of breast cancer in women living in Southwestern United States.
Breast cancer research and treatment (2006-10-20)
M L Slattery, C Sweeney, R Wolff, J Herrick, K Baumgartner, A Giuliano, T Byers
ABSTRACT
An insulin-related pathway to breast cancer has been hypothesized. We examine the 19 CA repeat of the IGF1 gene, the -202 C > A IGFBP3, the G972R IRS, and the G1057D IRS2 polymorphisms among 1,175 non-Hispanic white (NHW) and 576 Hispanic newly diagnosed breast cancer cases and 1,330 NHW and 727 Hispanic controls living in Arizona, Colorado, New Mexico, and Utah. Among post-menopausal women not recently exposed to hormones, not having the 19 CA repeat of IGF1 gene was associated with breast cancer among NHW women [odds ratio (OR) 2.14, 95% confidence interval (CI) 1.21-3.79] and having an R allele of G972R IRS1 increased breast cancer risk among Hispanic women (OR 2.70, 95% CI 1.13-6.46). Among post-menopausal Hispanic women recently exposed to hormones the A allele of the -202 C > A IGFBP3 polymorphism increased risk of breast cancer (OR 1.57, 95% CI 1.06-2.33). The IGF1 19 CA repeat polymorphism interacted with hormone replacement therapy (HRT) among NHW post-menopausal women; women who had the 19/19 IGF1 genotype were at reduced risk of breast cancer (OR 0.64, 95% CI 0.47-0.88) if they did not use HRT. We also observed interaction between body mass index and IGF1 19 CA repeat (p=0.06) and between weight gain and the -202 C > A IGFBP3 polymorphism (p=0.05) in NHW post-menopausal women not recently exposed to hormones. Our data suggest that associations between insulin-related genes and breast cancer risk among women living in the Southwestern United States may be dependent on estrogen exposure and may differ by ethnicity.
Sigma-Aldrich
IRS1 (600-1245), GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution