Merck
  • Silylarene Hydrogenation: A Strategic Approach that Enables Direct Access to Versatile Silylated Saturated Carbo- and Heterocycles.
Silylarene Hydrogenation: A Strategic Approach that Enables Direct Access to Versatile Silylated Saturated Carbo- and Heterocycles.
Angewandte Chemie (International ed. in English) (2018-05-24)
Mario P Wiesenfeldt, Tobias Knecht, Christoph Schlepphorst, Frank Glorius
ABSTRACT
We report a method to convert readily available silylated arenes into silylated saturated carbo- and heterocycles by arene hydrogenation. The scope includes alkoxy- and halosilyl substituents. Silyl groups can be derivatized into a plethora of functionalities and find application in organic synthesis, materials science, and pharmaceutical, agrochemical, and fragrance research. However, silylated saturated (hetero- ) cycles are difficult to access with current technologies. The yield of the hydrogenation depends on the amount of the silica gel additive. This silica effect also enables a significant improvement of a previously disclosed method for the hydrogenation of highly fluorinated arenes (e.g., to all-cis-C6 H6 F6 ).
Sigma-Aldrich
(CAAC-Cy)Rh(COD)Cl