Merck
  • Non-flammable electrolyte enables Li-metal batteries with aggressive cathode chemistries.
Non-flammable electrolyte enables Li-metal batteries with aggressive cathode chemistries.
Nature nanotechnology (2018-07-18)
Xiulin Fan, Long Chen, Oleg Borodin, Xiao Ji, Ji Chen, Singyuk Hou, Tao Deng, Jing Zheng, Chongyin Yang, Sz-Chian Liou, Khalil Amine, Kang Xu, Chunsheng Wang
ABSTRACT
Rechargeable Li-metal batteries using high-voltage cathodes can deliver the highest possible energy densities among all electrochemistries. However, the notorious reactivity of metallic lithium as well as the catalytic nature of high-voltage cathode materials largely prevents their practical application. Here, we report a non-flammable fluorinated electrolyte that supports the most aggressive and high-voltage cathodes in a Li-metal battery. Our battery shows high cycling stability, as evidenced by the efficiencies for Li-metal plating/stripping (99.2%) for a 5 V cathode LiCoPO4 (~99.81%) and a Ni-rich LiNi0.8Mn0.1Co0.1O2 cathode (~99.93%). At a loading of 2.0 mAh cm-2, our full cells retain ~93% of their original capacities after 1,000 cycles. Surface analyses and quantum chemistry calculations show that stabilization of these aggressive chemistries at extreme potentials is due to the formation of a several-nanometre-thick fluorinated interphase.
Sigma-Aldrich
Fluoroethylene carbonate, 99%
Sigma-Aldrich
1,1,2,2-Tetrafluoroethyl 2,2,2-trifluoroethyl ether, ≥99.5%, anhydrous, acid <=100 ppm, battery grade