Merck
  • Dissociated effect and Chemosensitive enhancement of tumor spheroids influenced by an electric field in a microdevice.
Dissociated effect and Chemosensitive enhancement of tumor spheroids influenced by an electric field in a microdevice.
Biomedical microdevices (2018-08-11)
Chun-Hao Huang, Kin Fong Lei, Ngan-Ming Tsang
ABSTRACT
The use of electric field for cancer therapy has been proposed for a novel non-invasive cancer therapeutic approach that provides better quality of life for patients. However, argument of the efficacy hampers the therapeutic development for various cancer diseases. More scientific evidences are necessary to be addressed by basic research. The current in vitro cell culture study reports the responses of tumor spheroids after the application of an alternating electric field. Human hepatocarchinoma cells suspended in soft hydrogel were cultured in a cell culture device embedded with stimulating electrodes. Tumor spheroids gradually formed and alternating electric field was then applied during the culture course. Investigation of cell viability and cell cycle were conducted to optimize the treatment conditions. The results showed that the electric potential of 1.0 Vpp and frequency of 130 kHz was the minimal effective conditions for inhibiting tumor spheroids. Importantly, dissociation of tumor spheroids was observed after the treatment. The effectiveness of chemotherapeutic agents was shown to be enhanced while the electric filed was simultaneously applied to the tumor spheroids. These results provided solid foundation for developing the effective therapeutic strategies.
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)