Merck
  • Glutamine-utilizing transaminases are a metabolic vulnerability of TAZ/YAP-activated cancer cells.
Glutamine-utilizing transaminases are a metabolic vulnerability of TAZ/YAP-activated cancer cells.
EMBO reports (2018-04-18)
Chih-Sheng Yang, Eleni Stampouloglou, Nathan M Kingston, Liye Zhang, Stefano Monti, Xaralabos Varelas
ABSTRACT
The transcriptional regulators TAZ and YAP (TAZ/YAP) have emerged as pro-tumorigenic factors that drive many oncogenic traits, including induction of cell growth, resistance to cell death, and activation of processes that promote migration and invasion. Here, we report that TAZ/YAP reprogram cellular energetics to promote the dependence of breast cancer cell growth on exogenous glutamine. Rescue experiments with glutamine-derived metabolites suggest an essential role for glutamate and α-ketoglutarate (AKG) in TAZ/YAP-driven cell growth in the absence of glutamine. Analysis of enzymes that mediate the conversion of glutamate to AKG shows that TAZ/YAP induce glutamic-oxaloacetic transaminase (GOT1) and phosphoserine aminotransferase (PSAT1) expression and that TAZ/YAP activity positively correlates with transaminase expression in breast cancer patients. Notably, we find that the transaminase inhibitor aminooxyacetate (AOA) represses cell growth in a TAZ/YAP-dependent manner, identifying transamination as a potential vulnerable metabolic requirement for TAZ/YAP-driven breast cancer.
Sigma-Aldrich
Dimethyl 2-oxoglutarate, 96%
Sigma-Aldrich
O-(Carboxymethyl)hydroxylamine hemihydrochloride, 98%
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
L-Serine methyl ester hydrochloride, 98%
Sigma-Aldrich
BPTES, ≥95% (HPLC)