Merck
  • The Hepatitis Delta Virus accumulation requires paraspeckle components and affects NEAT1 level and PSP1 localization.
The Hepatitis Delta Virus accumulation requires paraspeckle components and affects NEAT1 level and PSP1 localization.
Scientific reports (2018-04-18)
Yasnee Beeharry, Gabrielle Goodrum, Christian J Imperiale, Martin Pelchat
ABSTRACT
The Hepatitis Delta Virus (HDV) relies mainly on host proteins for its replication. We previously identified that PSF and p54nrb associate with the HDV RNA genome during viral replication. Together with PSP1, these proteins are part of paraspeckles, which are subnuclear bodies nucleated by the long non-coding RNA NEAT1. In this work, we established the requirement for PSF, p54nrb and PSP1 in HDV replication using RNAi-mediated knockdown in HEK-293 cells replicating the HDV RNA genome. We determined that HDV replication induces the delocalization of PSP1 to cytoplasmic foci containing PABP and increases NEAT1 level causing an enlargement of NEAT1 foci. Overall, our data support a role for the main paraspeckles proteins in HDV life cycle and indicate that HDV replication causes a cellular stress and induces both a delocalization of the PSP1 to the cytoplasm and a disruption of paraspeckles.
Sigma-Aldrich
Anti-PSPC1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-p54nrb/NonO Antibody, clone 78-1-C6, clone 78-1-C6, Upstate®, from mouse