Zileuton
Synonym(s):
(±)-N-hydroxy-N-(1-benzo[b]thien-2-ylethyl)urea, Zileuton
Empirical Formula (Hill Notation):
C11H12N2O2S
CAS Number:
Molecular Weight:
236.29
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing