![Violet Red Bile Glucose Agar suitable for microbiology, NutriSelect® Plus](/deepweb/assets/sigmaaldrich/product/images/389/853/4144eec7-7898-4b0e-835a-41627c91d1de/640/4144eec7-7898-4b0e-835a-41627c91d1de.jpg)
Violet Red Bile Glucose Agar
![Violet Red Bile Glucose Agar suitable for microbiology, NutriSelect® Plus](/deepweb/assets/sigmaaldrich/product/images/389/853/4144eec7-7898-4b0e-835a-41627c91d1de/640/4144eec7-7898-4b0e-835a-41627c91d1de.jpg)
Synonym(s):
VRBG-Agar
Products
Product Category
Brand
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing