Valaciclovir impurity F para-toluenesulfonate
Synonym(s):
L-Valine 2-hydroxyethyl ester p-toluenesulfonate salt
Empirical Formula (Hill Notation):
C7H15NO3 · C7H8O3S
CAS Number:
Molecular Weight:
333.40
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing