![Valaciclovir impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/160/429/6e72e657-3546-405b-90a0-4160b455c749/640/6e72e657-3546-405b-90a0-4160b455c749.png)
Valaciclovir impurity E
![Valaciclovir impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/160/429/6e72e657-3546-405b-90a0-4160b455c749/640/6e72e657-3546-405b-90a0-4160b455c749.png)
Synonym(s):
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-(benzyloxycarbonyl)-L-valinate, N-[(Phenylmethoxy)carbonyl]-L-valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester
Empirical Formula (Hill Notation):
C21H26N6O6
CAS Number:
Molecular Weight:
458.47
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing