TSKgel® G2000SW (10 µm) HPLC Columns
Synonym(s):
TSKgel® Size Exclusion (SW-Type) HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2