
Trimetazidine for system suitability

Synonym(s):
1-(2,3,4-Trimethoxybenzyl)piperazine dihydrochloride, Trimetazidine dihydrochloride
Empirical Formula (Hill Notation):
C14H22N2O3 · 2HCl
CAS Number:
Molecular Weight:
339.26
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing