![trans,trans-Farnesyl pyrophosphate ammonium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/170/934/88eae626-ef0a-4e3f-b6c2-083aa3ef306b/640/88eae626-ef0a-4e3f-b6c2-083aa3ef306b.png)
trans,trans-Farnesyl pyrophosphate ammonium salt
![trans,trans-Farnesyl pyrophosphate ammonium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/170/934/88eae626-ef0a-4e3f-b6c2-083aa3ef306b/640/88eae626-ef0a-4e3f-b6c2-083aa3ef306b.png)
Synonym(s):
(E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl pyrophosphate ammonium salt, Farnesyl diphosphate ammonium salt
Empirical Formula (Hill Notation):
C15H28O7P2 · xNH3
Molecular Weight:
382.33 (free acid basis)
Products
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing