trans-o-Coumaric acid
Synonym(s):
trans-2-Hydroxycinnamic acid, (E)-3-(2-Hydroxyphenyl)acrylic acid, o-Coumaric acid
Empirical Formula (Hill Notation):
C9H8O3
CAS Number:
Molecular Weight:
164.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing