![Transforming Growth Factor-β3 human TGF-β3, recombinant, expressed in baculovirus infected Sf21 cells, powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/206/208/9cda0a4c-cf91-495f-a4f7-f4aceb61650e/640/9cda0a4c-cf91-495f-a4f7-f4aceb61650e.jpg)
Transforming Growth Factor-β3 human
![Transforming Growth Factor-β3 human TGF-β3, recombinant, expressed in baculovirus infected Sf21 cells, powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/206/208/9cda0a4c-cf91-495f-a4f7-f4aceb61650e/640/9cda0a4c-cf91-495f-a4f7-f4aceb61650e.jpg)
Synonym(s):
TGF-β3
Products
Product Category
Brand
Biological Source
Gene Alias
Manufacturer
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing