![trans-Dichlorobis(triphenylphosphine)platinum(II)](/deepweb/assets/sigmaaldrich/product/structures/146/545/8f9148ed-7de9-4be6-adbd-ef3424d3b0ef/640/8f9148ed-7de9-4be6-adbd-ef3424d3b0ef.png)
trans-Dichlorobis(triphenylphosphine)platinum(II)
![trans-Dichlorobis(triphenylphosphine)platinum(II)](/deepweb/assets/sigmaaldrich/product/structures/146/545/8f9148ed-7de9-4be6-adbd-ef3424d3b0ef/640/8f9148ed-7de9-4be6-adbd-ef3424d3b0ef.png)
Synonym(s):
trans-Bis(triphenylphosphine)platinum dichloride
Linear Formula:
[(C6H5)3P]2PtCl2
CAS Number:
Molecular Weight:
790.56
Products
Brand
Formula Weight
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing