Skip to Content
Merck
TopFluor™-SOBRAC powder, Avanti Research™ - A Croda Brand

TopFluor-SOBRAC

TopFluor™-SOBRAC powder, Avanti Research™ - A Croda Brand
Synonym(s):
TopFluor-SOBRAC
Empirical Formula (Hill Notation):
C28H41BBrF2N3O3
CAS Number:
Molecular Weight:
596.36

Products

Formula Weight
Manufacturer
Physical Form
Purity
Application
Package Size
Showing 1-2 of 2