![Terbufos-sulfone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/368/287/ba9b5f68-3d51-4cd2-b213-a4efcf7011bf/640/ba9b5f68-3d51-4cd2-b213-a4efcf7011bf.png)
Terbufos-sulfone
![Terbufos-sulfone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/368/287/ba9b5f68-3d51-4cd2-b213-a4efcf7011bf/640/ba9b5f68-3d51-4cd2-b213-a4efcf7011bf.png)
Empirical Formula (Hill Notation):
C9H21O4PS3
CAS Number:
Molecular Weight:
320.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing