
tBuXPhos Pd G1

Synonym(s):
t-BuXPhos palladium(II) phenethylamine chloride, tBuXPhos-Pd-G1, Chloro[2-(di-tert-butylphosphino)-2′,4′,6′-triisopropyl-1,1′-biphenyl][2-(2-aminoethyl)phenyl)]palladium(II), [2-(Di-tert-butylphosphino)-2′,4′,6′-triisopropyl-1,1′-biphenyl][2-(2-aminoethyl)phenyl)]palladium(II) chloride, t-BuXPhos Palladacycle, t-BuXPhos precatalyst
Empirical Formula (Hill Notation):
C37H55ClNPPd
CAS Number:
Molecular Weight:
686.69
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing