Skip to Content
Merck
SUPELCOSIL™ LC-18 HPLC Column 5 μm particle size, L × I.D. 5 cm × 4 mm

SUPELCOSIL LC-18 (5 µm) HPLC Columns

SUPELCOSIL™ LC-18 HPLC Column 5 μm particle size, L × I.D. 5 cm × 4 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing