Skip to Content
Merck
vibrant-m

Sulbactam Related Compound E

vibrant-m
Synonym(s):
(2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, 6,6-Dibromopenicillanic acid 1,1-dioxide
Empirical Formula (Hill Notation):
C8H9Br2NO5S
CAS Number:
Molecular Weight:
391.03
EC Number:

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1