Skip to Content
Merck
Sucrose-(glucose-13C6) endotoxin tested, ≥99 atom % 13C, ≥99% (CP)

Sucrose-(glucose-13C6)

Sucrose-(glucose-13C6) endotoxin tested, ≥99 atom % 13C, ≥99% (CP)
Synonym(s):
D-(+)-Saccharose-(glucose-13C6), Sugar-(glucose-13C6)
Empirical Formula (Hill Notation):
13C6C6H22O11
CAS Number:
Molecular Weight:
348.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Optical Isomer
Showing 1-2 of 2