
Sodium phenylbutyrate impurity C

Synonym(s):
Cyclohexanebutyric acid, 4-Cyclohexylbutyric acid
Linear Formula:
C6H11(CH2)3CO2H
CAS Number:
Molecular Weight:
170.25
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing