Skip to Content
Merck
Sodium hexanitrocobaltate(III) ACS reagent

Sodium hexanitrocobaltate(III)

Sodium hexanitrocobaltate(III) ACS reagent
Synonym(s):
Sodium cobaltnitrite, Sodium hexanitritocobaltate(III), Sodium hexanitrocobaltate(III)
Linear Formula:
Na3[Co(NO2)6]
CAS Number:
Molecular Weight:
403.94

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing