Serine
Synonym(s):
L-Serine, (S)-2-Amino-3-hydroxypropionic acid
Linear Formula:
HOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
105.09
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing