Skip to Content
Merck
S-(4-Nitrobenzyl)-6-thioinosine ≥98%, solid

S-(4-Nitrobenzyl)-6-thioinosine

S-(4-Nitrobenzyl)-6-thioinosine ≥98%, solid
Synonym(s):
6-[(4-Nitrobenzyl)thio]-9-β-D-ribofuranosylpurine, NBMPR, NBTI
Empirical Formula (Hill Notation):
C17H17N5O6S
CAS Number:
Molecular Weight:
419.41
EC Number:

Products

Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing