Skip to Content
Merck
(R)-(−)-sec-Butylamine 99%

(R)-(−)-sec-Butylamine

(R)-(−)-sec-Butylamine 99%
Synonym(s):
(R)-(−)-2-Aminobutane
Linear Formula:
C2H5CH(NH2)CH3
CAS Number:
Molecular Weight:
73.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing