(R)-(+)-8-Hydroxy-DPAT hydrobromide
Synonym(s):
(R)-(+)-2-Dipropylamino-8-hydroxy-1,2,3,4-tetrahydronaphthalene hydrobromide, (R)-(+)-8-Hydroxy-2-(dipropylamino)tetralin hydrobromide
Empirical Formula (Hill Notation):
C16H25NO · HBr
CAS Number:
Molecular Weight:
328.29
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing