Skip to Content
Merck
Propidium iodide ≥94% (HPLC)

Propidium iodide

Propidium iodide ≥94% (HPLC)
Synonym(s):
3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, Propidium Iodide
Empirical Formula (Hill Notation):
C27H34I2N4
CAS Number:
Molecular Weight:
668.39
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing