![Prilocaine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/180/342/766c0da9-a2bc-4bcc-9d72-a2b556a7c014/640/766c0da9-a2bc-4bcc-9d72-a2b556a7c014.png)
Prilocaine impurity B
![Prilocaine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/180/342/766c0da9-a2bc-4bcc-9d72-a2b556a7c014/640/766c0da9-a2bc-4bcc-9d72-a2b556a7c014.png)
Synonym(s):
o-Toluidine hydrochloride, 2-Methylaniline hydrochloride
Linear Formula:
CH3C6H4NH2 · HCl
CAS Number:
Molecular Weight:
143.61
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing