![Pinosylvin monomethyl ether phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/299/136/ba1debb3-fd8a-4bab-b795-5275ea3491ca/640/ba1debb3-fd8a-4bab-b795-5275ea3491ca.png)
Pinosylvin monomethyl ether
![Pinosylvin monomethyl ether phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/299/136/ba1debb3-fd8a-4bab-b795-5275ea3491ca/640/ba1debb3-fd8a-4bab-b795-5275ea3491ca.png)
Synonym(s):
(E)-3-Hydroxy-5-methoxystilbene, (E)-3-Methoxy-5-(2-phenylethenyl)phenol, 5-Methoxy-3-stilbenol, Pinosylvin 3-methyl ether
Empirical Formula (Hill Notation):
C15H14O2
CAS Number:
Molecular Weight:
226.27
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing