Skip to Content
Merck
Phosphoramidon disodium salt ≥97% (HPLC)

Phosphoramidon disodium salt

Phosphoramidon disodium salt ≥97% (HPLC)
Synonym(s):
N-(α-Rhamno­pyranosyl­oxy­hydroxy­phosphinyl)-Leu-Trp disodium salt, N-(α-Rhamno­pyranosyl­phos­phono)-L-leucyl-L-tryptophan disodium salt
Empirical Formula (Hill Notation):
C23H32N3Na2O10P
CAS Number:
Molecular Weight:
587.47

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing