Skip to Content
Merck
Phencyclidine hydrochloride

Phencyclidine hydrochloride

Phencyclidine hydrochloride
Synonym(s):
1-(1-Phenylcyclohexyl)piperidine hydrochloride, PCP hydrochloride
Empirical Formula (Hill Notation):
C17H25N · HCl
CAS Number:
Molecular Weight:
279.85

Products

Product Category
Brand
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing