Skip to Content
Merck
Oxytetracycline for system suitability A European Pharmacopoeia (EP) Reference Standard

Oxytetracycline for system suitability A

Oxytetracycline for system suitability A European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Oxytetracycline
Empirical Formula (Hill Notation):
C22H24N2O9
CAS Number:
Molecular Weight:
460.43

Products

Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1