Oxydemeton-methyl
Synonym(s):
Demeton-S-methyl sulfoxide, S-[2-(Ethylsulfinyl)ethyl] O,O-dimethyl phosphorothioate, Methyl oxydemeton
Empirical Formula (Hill Notation):
C6H15O4PS2
CAS Number:
Molecular Weight:
246.28
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing