![Oxasulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/571/873/054d2cbc-d236-44eb-b7a1-139a7b036290/640/054d2cbc-d236-44eb-b7a1-139a7b036290.png)
Oxasulfuron
![Oxasulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/571/873/054d2cbc-d236-44eb-b7a1-139a7b036290/640/054d2cbc-d236-44eb-b7a1-139a7b036290.png)
Synonym(s):
Oxetan-3-yl 2-[(4,6-dimethylpyrimidin-2-yl)carbamoylsulfamoyl]benzoate
Empirical Formula (Hill Notation):
C17H18N4O6S
CAS Number:
Molecular Weight:
406.41
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing