ORBO™ 32 Small Activated Coconut Charcoal (20/40), 100/50 mg
Synonym(s):
Activated Coconut Charcoal Tube
Products
Brand
Product Line
Agency
Chemical Composition
OD (mm)
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing