Skip to Content
Merck
Omeprazole Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material

Omeprazole Related Compound E

Omeprazole Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Omeprazole N-oxide, 4-Methoxy-2-{[(RS)-(5-methoxy-1H-benzimidazol-2-yl)sulfinyl]methyl}-3,5-dimethylpyridine 1-oxide
Empirical Formula (Hill Notation):
C17H19N3O4S
CAS Number:
Molecular Weight:
361.42

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique