Skip to Content
Merck
Oleoyl-L-α-lysophosphatidic acid sodium salt ≥98%, solid

Oleoyl-L-α-lysophosphatidic acid sodium salt

Oleoyl-L-α-lysophosphatidic acid sodium salt ≥98%, solid
Synonym(s):
1-Oleoyl-sn-glycerol 3-phosphate sodium salt, 3-sn-Lysophosphatidic acid, 1-oleoyl sodium salt, LPA sodium salt
Empirical Formula (Hill Notation):
C21H41O7P · xNa+
CAS Number:
Molecular Weight:
436.52 (free acid basis)

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing