Skip to Content
Merck
Oleic acid-13C18 ≥99 atom % 13C, ≥99% (CP)

Oleic acid-13C18

Oleic acid-13C18 ≥99 atom % 13C, ≥99% (CP)
Synonym(s):
13C Labeled oleic acid, Elainic acid-13C18, Oleic acid-ul-13C
Linear Formula:
13CH3(13CH2)713CH=13CH(13CH2)713CO2H
CAS Number:
Molecular Weight:
300.33

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing